missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DreamTaq™ Hot Start DNA Polymerase

DNA Polymerase is the product of choice for consistently robust amplification with minimal optimization steps

Brand:  DreamTaq EP1703

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15797496

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Includes: DreamTaq Hot Start DNA Polymerase; 10X DreamTaq Buffer

Description

Description

Thermo Scientific™ DreamTaq™ Hot Start DNA Polymerase is an enhanced hot start Taq DNA polymerase that ensures higher PCR specificity, sensitivity and yields compared to conventional hot start Taq DNA polymerases. DreamTaq Hot Start DNA Polymerase uses antibody based inhibition of DNA polymerase activity at ambient temperatures, thus preventing the amplification of non-specific products prior to the amplification step.

Better reaction outcomes:
  • Higher yield of target amplicons from low template amounts.
  • Increased reaction specificity due to reliable hot-start technology.
  • Wider range of amplicon lengths - routine amplification of genomic DNA fragments up to 6kb.
Enhanced convenience:
  • Room temperature reaction set-up.
  • Minimized optimization of Mg2+ concentration and of primer annealing temperatures due to optimized reaction buffer.
Amplicon size (complex DNA): 6 kb
Fidelity (vs. Taq): 1X

Recommended Storage

Store at -20C

Specifications

Specifications

Contains:
• 5 x 100 μL DreamTaq Hot Start DNA Polymerase (5 U/μL)
• 10 x 1.25 mL 10X DreamTaq Buffer (includes 20 mM MgCl2)

Store at -5 to -30°C.
DreamTaq DNA Polymerase
DreamTaq™
Wet or Dry Ice
3'-A
Low
Standard
2500 U
Built-In Hot Start
Separate Components
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers