missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente™ Drechsel Gas Washing Bottles

Gas washing bottles acc. to Drechsel with filter disc

£81.80 - £120.00

Specifications

Includes DURAN™ tubing head, borosilicate glass 3.3 bottle
View More Specs

Products 3
Product Code Brand Capacity (Metric) Porosity Price Quantity & Availability  
Product Code Brand Capacity (Metric) Porosity Price Quantity & Availability  
11536924
View Documents
Lenz Laborglasintrumente™
05510113
500 mL 1
£120.00
Each
Estimated Shipment: 28-05-2025
to see stock.
Add to basket
 
10450323
View Documents
Lenz Laborglasintrumente™
05510258
500 mL 2
£119.60
Each
In Stock
to see stock.
Add to basket
 
10622092
View Documents
Lenz Laborglasintrumente™
05510270
1000 mL 2
£81.80
Each
Estimated Shipment: 21-05-2025
to see stock.
Add to basket
 
Description

Description

  • Acc. to DIN 12596
  • Complete with head (with filter disc)
  • Ground joint connection: NS 29/32
  • Material: DURAN™ tubing (head), borosilicate glass 3.3 (bottle)
Specifications

Specifications

DURAN™ tubing head, borosilicate glass 3.3 bottle
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lenz Laborglasintrumente™ Drechsel Gas Washing Bottles

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.