missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DRI™ Endocrine Assays

Achieve excellent workflow management, cost savings and superior performance with DRI Endocrine Assays

£1350.00

Specifications

Content And Storage 2°C to 8°C
Sample Type Plasma, Serum
View More Specs

Products 1
Product Code Brand Quantity Type Price Quantity & Availability  
Product Code Brand Quantity Type Price Quantity & Availability  
17730308
View Documents
Thermo Scientific™
0454
100 mL, 34 mL Total Thyroxine Assay
£1350.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Achieve excellent workflow management, cost savings and superior performance with Thermo Scientific™ DRI™ Endocrine Assays, packaged in convenient sizes with applications on a multitude of clinical chemistry analyzers. Endocrine Assays T4 and T Uptake are available in both DRI and Thermo Scientific™ CEDIA™ formats.

Specifications

Specifications

2°C to 8°C
Plasma, Serum
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ DRI™ Endocrine Assays

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.