missing translation for 'onlineSavingsMsg'
Learn More

Drierite™, with indicator, 10-20 mesh, Thermo Scientific Chemicals

£52.86 - £526.62

Chemical Identifiers

CAS 7778-18-9, 7646-79-9
Molecular Formula CaO4S
Molecular Weight (g/mol) 136.13
MDL Number MFCD00010912
InChI Key OSGAYBCDTDRGGQ-UHFFFAOYSA-L
PubChem CID 24497
ChEBI CHEBI:31346
SMILES [Ca++].[O-]S([O-])(=O)=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10408862
View Documents
Thermo Scientific Acros
350015000
500 g Glass Bottle
£52.86
500g
Please to purchase this item. Need a web account? Register with us today!
10366781
View Documents
Thermo Scientific Acros
350010020
2 kg Glass Bottle
£154.77
2kg
Please to purchase this item. Need a web account? Register with us today!
11984251
View Documents
Thermo Scientific Acros
350010100
10 kg Plastic Drum
£526.62
10kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent, Indicator
Specifications

Chemical Identifiers

7778-18-9, 7646-79-9
136.13
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
CHEBI:31346
CaO4S
MFCD00010912
24497
[Ca++].[O-]S([O-])(=O)=O

Specifications

Blue
Irregular Granules
CaO4S
3077
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
[Ca++].[O-]S([O-])(=O)=O
136.13
CHEBI:31346
Drierite™
1450.0°C
7778-18-9, 7646-79-9
MFCD00010912
Solubility in water: slightly soluble. Other solubilities: slightly soluble in glycerol, practically insoluble in most common organic, solvents. SiO2 and insolubles: 0.58%
0.29%
calcium sulfate
24497
136.13
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Suspected of causing genetic defects. May cause cancer by inhalation. May damage fertility. Causes serious eye irritation. Toxic to aquatic life with long lasting effects.

GHS P Statement
Obtain special instructions before use. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection. If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician. IF exposed or concerned: Get medical advice/attention.

Danger

Product Identifier
  • Drierite, with indicator

Signal Word
  • Danger

Hazard Category
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 1B
  • LONG-TERM AQUATIC HAZARD Chronic 2

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contains : Calcium sulfate, Cobalt(II) chloride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only