missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Drierite, with indicator, 8 mesh, Thermo Scientific Chemicals
138.00 GBP valid until 2024-07-31
Use promo code "23251" to get your promotional price.
Use promo code "23251" to get your promotional price.
Click to view available options
Quantity:
500 g
2 kg
5 kg
Packaging:
Glass Bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent, Indicator
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Blue |
Physical Form | Granular |
Quantity | 2 kg |
Fieser | 01,107 |
Merck Index | 15, 1708 |
Packaging | Glass Bottle |
Solubility Information | SiO2 and insolubles: 0.58% |
R2O | 0.29% |
IUPAC Name | calcium;sulfate |
Formula Weight | 136.13 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS09_Aquatic_Toxicity.png)
Product Identifier
- Drierite, with indicator
- Danger
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1B
- Reproductive toxicity Category 1B
- LONG-TERM AQUATIC HAZARD Chronic 2
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H341-Suspected of causing genetic defects.
- H350i-May cause cancer by inhalation.
- H360F-May damage fertility.
- H411-Toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contains : Cobalt(II) chloride
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'