missing translation for 'onlineSavingsMsg'
Learn More

Drierite™, without indicator, 10-20 mesh, Thermo Scientific Chemicals

£40.90 - £444.00

Chemical Identifiers

CAS 7778-18-9
Molecular Formula CaO4S
Molecular Weight (g/mol) 136.13
MDL Number MFCD00010912
InChI Key OSGAYBCDTDRGGQ-UHFFFAOYSA-L
PubChem CID 24497
ChEBI CHEBI:31346
SMILES [Ca++].[O-]S([O-])(=O)=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10472601
View Documents
Thermo Scientific Acros
219065000
500 g Glass Bottle
£40.90
500g
Please to purchase this item. Need a web account? Register with us today!
10348020
View Documents
Thermo Scientific Acros
219060020
2 kg Plastic Bottle
£116.00
2kg
Please to purchase this item. Need a web account? Register with us today!
11964251
View Documents
Thermo Scientific Acros
219060100
10 kg Plastic Drum
£444.00
10kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent, Indicator
Specifications

Chemical Identifiers

7778-18-9
136.13
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
CHEBI:31346
CaO4S
MFCD00010912
24497
[Ca++].[O-]S([O-])(=O)=O

Specifications

1450.0°C
7778-18-9
MFCD00010912
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
calcium;sulfate
24497
136.14
Granular
CaO4S
Solubility in water: ca. 0.25g/L (20°C). Other solubilities: insoluble in organic liquids, slightly soluble in glycerol
[Ca++].[O-]S([O-])(=O)=O
136.13
CHEBI:31346
Drierite™
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Drierite]TS, without indicator

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only