missing translation for 'onlineSavingsMsg'
Learn More

Drierite™, without indicator, 6 mesh

Product Code. 10452411 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 g
2 kg
10 kg
Packaging:
Glass Bottle
Plastic Drum
Plastic bottle
Unit Size:
10kg
2kg
500g
This item is not returnable. View return policy

Product Code. 10452411

Brand: Thermo Scientific Acros 219045000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7778-18-9 | CaO4S | 136.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent, Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-18-9
Molecular Formula CaO4S
Molecular Weight (g/mol) 136.13
MDL Number MFCD00010912
InChI Key OSGAYBCDTDRGGQ-UHFFFAOYSA-L
PubChem CID 24497
ChEBI CHEBI:31346
SMILES [Ca++].[O-]S([O-])(=O)=O

Specifications

Melting Point 1450.0°C
Physical Form Granular
Quantity 500 g
Packaging Glass Bottle
Solubility Information Solubility in water: ca. 0.25g/L (20°C). Other solubilities: insoluble in organic liquids, slightly soluble in glycerol
IUPAC Name calcium sulfate
Formula Weight 136.14
Chemical Name or Material Drierite™

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.