Learn More
Drinking Water Standards without Mercury for Environmental Analysis, Set of 2, SPEX CertiPrep™
Drinking Water Pollutant Standards without Mercury for AA & ICP, 125 mL, SPEX CertiPrep™. ISO Guide 34 Certified, NIST Traceable Certified Reference Materials. Supplied with comprehensive Certificate of Analysis.
Brand: SPEX Certiprep DW-SETN
Code : NEW
Additional Details : Weight : 0.09000kg Transport : UN number : 3264 Chem class : 8 Pack group : III
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
- Drinking Water Standards
- 125 mL volume
- Includes EP-8N and EP-4
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Compliance
GHS/OSHA Complaint
Specifications
Drinking Water Standards without Mercury | |
2 x 125 mL | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
2% HNO3 |
Triple-leached LDPE bottle | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Environmental |
Product intended for laboratory use only