Learn More
Thermo Scientific™ μDrop™ and μDrop Duo Plates
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ μDrop™ and μDrop™ Duo Plates are ideal tools for quick and easy photometric low-volume (2–10 μl) concentration measurements of DNA, RNA, protein samples, or spectral scanning. The low volume area is designed to be used with 8-channel pipette for fast and accurate multiple sample pipetting. The plates also contain a separate area for cuvette measurement. μDrop Plates are compatible with Thermo Scientific Varioskan™ LUX, Multiskan™ Sky and Multiskan SkyHigh microplate readers. The intuitive Thermo Scientific SkanIt™ software and Multiskan SkyHigh user interface offer ready-made measurement protocols for quick sample measurement and analysis.
- dsDNA detection range from a few micrograms to a few milligrams per milliliter
- Up to 16 (μDrop Plate) or 32 (μDrop Duo Plate) sample positions for quick and easy measurement of sample volumes down to 2 μl
- Compatible with an eight-channel pipette for sample addition
- Easy to clean after measurement
- Measurement slot for a rectangular cuvette
- Ready-made Multiskan SkyHigh user interface and SkanIt Software protocols for nucleic acid and protein analysis
- Compatible with Multiskan Sky, Multiskan SkyHigh and Varioskan LUX microplate readers
Specifications
Specifications
| Product Type | μDrop Duo Plate |
| For Use With (Equipment) | Varioskan LUX, Multiskan Sky, Multiskan SkyHigh |
| Plate Type | μDrop Duo Plate |
| Capacity (Metric) | 2 to 10μL (low-volume function) |
| For Use With (Application) | Nucleic acid and protein concentration measurement |
| No. of Positions | 32 |