Learn More
DWK Life Sciences Dropping Bottle, Amber, Soda-Lime Glass, Supplied with ground jointed pipette, Complete with Rubber teat
Shop All DWK Life Sciences ProductsDescription
Dropping bottles are suitable for liquids which need to be dispensed in small amounts, especially drop by drop. Amber glass bottle provides protection from light for photosensitive reagents. Supplied with rubber teat and clear ground jointed dropping pipette. Replacement clear glass pipettes and natural rubber teats are available.
- Amber glass bottle provides protection from light for photosensitive reagents
- Clear glass pipette with standard ground joint
- Tight closure due to standard taper ground joint
- Natural rubber teat

Specifications
Specifications
Capacity (Metric) | 50 mL |
Description | From borosilicate 3.3 |
For Use With (Application) | Storage and dispensing of photosensitive liquids and reagents |
Material | Soda Lime Glass |
Quantity | 10/Pk. |
Autoclavable | Not Autoclavable |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
New Offer Save Now -
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70% -
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.