Learn More
Lenz Laborglasinstrumente™ Dropping funnel, conical-shaped with glass plug
Made from borosilicate glass. Lenz Laborglasinstrumente™ Dropping funnel, conical-shaped with glass plug is used in the laboratory to transfer fluids. The funnel is fitted with a stopcock which allows the flow to be controlled.
£72.70 - £187.00
Specifications
Material Borosilicate Glass
View More Specs
Products 4
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
11876074
View Documents
Lenz Laborglasinstrumente™
04452802
50 mL
£72.70
Each
Estimated Shipment: 20-06-2025
to see stock.
Add to basket
11866124
View Documents
Lenz Laborglasinstrumente™
04455804
500 mL
£89.90
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
11876114
View Documents
Lenz Laborglasinstrumente™
04457004
1000 mL
£111.00
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
11886144
View Documents
Lenz Laborglasinstrumente™
04457304
2000 mL
£187.00
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
Description
Description
  • Conical shape with NS socket and NS cone
  • With solid glass plug, with screwthread retaining device
  • With inscription label, without scale
  • Borosilicate glass
Specifications
Specifications
Borosilicate Glass
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Dropping funnel, conical-shaped with glass plug