missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Drosophila Bottles
Wide base/narrow top design prevents fly meal from falling out when bottle is turned upside down
£182.00
Specifications
Color | Transparent |
---|---|
Type | Stock Bottle |
For Use With (Application) | Straight wall design minimizes the dislodging of food, provides more efficient racking, filling and plugging |
Product Code | Brand | Bottom Shape | Capacity (English) | Material | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Bottom Shape | Capacity (English) | Material | Price | Quantity & Availability | |||
10590143
|
Fisherbrand™
|
Round | 6 oz. | Polypropylene |
£182.00
Pack of 500 |
Estimated Shipment: 29-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Wide base/narrow top design prevents fly meal from falling out when bottle is turned upside down
Drosophila bottles are available in transparent, autoclavable polypropylene and non-autoclavable polyethylene. The bottle openings are designed to accept standard paper lids, foam plugs, cotton balls and Buzz Plugs.
Round-Bottom Bottles
- Polypropylene
- Single use or autoclave for reuse
- Available in polypropylene or polyethylene
- Polypropylene provides good transparency and can be autoclaved for reuse or disposed of after single use
Specifications
Transparent | |
Stock Bottle | |
Straight wall design minimizes the dislodging of food, provides more efficient racking, filling and plugging |