missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRP1 (C-terminal) Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12957-1-AP-150UL
This item is not returnable.
View return policy
Description
Dynamin-related protein 1 (DRP1) is a member of the dynamin superfamily of GTPases. Members of this family have diverse cellular functions including vesicle scission, organelle fission, viral resistance, and intracellular trafficking (reviewed in 1). DRP1 affects mitochondrial morphology and is important in mitochondrial and peroxisomal fission in mammalian cells.Specifications
DRP1 (C-terminal) | |
Polyclonal | |
Unconjugated | |
DNM1L | |
DLP1, DNM1L, DRP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10059, 74006 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00429, Q8K1M6 | |
Dnm1l | |
DRP1 (C-terminal) Fusion Protein Ag3644 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |