missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Drum Rack With Sump Pallet PE

The Drum Rack With Sump Pallet PE provides safe storage and filling storage of substances hazardous to water of all hazard classes.

Brand:  asecos™ 13244

 View more versions of this product

Product Code. 17806516

  • £540.00 / Each
Estimated Shipment: 04-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For the storage of acids, alkalis and further aggressiv chemicals
  • Safe dispensing due to overhanging collecting sump
Specifications

Specifications

15.9 gal.
Steel
2972 in.
29.72 in.
61.81 in.
31.30 in.
92.59 lb.
60 L
Drum Rack
755.00 mm
755.00 mm
1570.00 mm
795.00 mm
42 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers