missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Drum Rack With Sump Pallet PE

The Drum Rack With Sump Pallet PE provides safe storage and filling storage of substances hazardous to water of all hazard classes.

Brand:  asecos™ 13137

 View more versions of this product

Product Code. 17816416

  • £748.00 / Each
Estimated Shipment: 04-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For the storage of acids, alkalis and further aggressiv chemicals
  • Safe dispensing due to overhanging collecting sump
Specifications

Specifications

47.6 gal.
Steel
52.36 in.
52.36 in.
61.81 in.
45.98 in.
167.55 lb.
180 L
Drum Rack
1330.00 mm
1330.00 mm
1570.00 mm
1168.00 mm
76 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers