Learn More
Thermo Scientific™ Drums for Digital Cel-Gro Tissue Culture Rotators
Accessory for use with Digital Cel-Gro Tissue Culture Rotator, 120V and Digital Cel-Gro Tissue Culture Rotator, 230V
Brand:  Thermo Scientific™ 88882152
Additional Details : Weight : 1.05000kg
 View more versions of this product
Product Code. 17308282
  • £149.72 / Each
Estimated Shipment: 07-11-2024
to see stock.
Add to basket
Description
Description
Accessory for use with Digital Cel-Gro Tissue Culture Rotator, 120V and Digital Cel-Gro Tissue Culture Rotator, 230V
Specifications
Specifications
Rotating Drum
290 mm
10/15 mL, 5/7 mL tubes
30
Stainless Steel
10 x 10/15 mL and 20 x 5/7 mL rotator drum
Accessory for Digital Cel-Gro Tissue Culture Rotator, 120V (88882015) and Digital Cel-Gro Tissue Culture Rotator 230V (88882016)