Learn More
Thermo Scientific™ Drums for Digital Cel-Gro Tissue Culture Rotators
Accessory for use with Digital Cel-Gro Tissue Culture Rotator, 120V and Digital Cel-Gro Tissue Culture Rotator, 230V
Brand:  Thermo Scientific™ 88882153
Additional Details : Weight : 1.50000kg
 View more versions of this product
Product Code. 17318282
  • £149.72 / Each
In Stock
to see stock.
Add to basket
Description
Description
Accessory for use with Digital Cel-Gro Tissue Culture Rotator, 120V and Digital Cel-Gro Tissue Culture Rotator, 230V
Specifications
Specifications
Rotating Drum
64 x diameter 14 mm tube rotator drum
64
310 mm
diameter 14 mm tube