Learn More
Thermo Scientific™ Drums for Digital Cel-Gro Tissue Culture Rotators
Accessory for use with Digital Cel-Gro Tissue Culture Rotator, 120V and Digital Cel-Gro Tissue Culture Rotator, 230V
Brand: Thermo Scientific™ 88882154
Additional Details : Weight : 1.55000kg
Description
Accessory for use with Digital Cel-Gro Tissue Culture Rotator, 120V and Digital Cel-Gro Tissue Culture Rotator, 230V
Specifications
Rotating Drum | |
310 mm | |
diameter 18.5 mm tube | |
64 |
Stainless Steel | |
64 x diameter 18.5 mm tube rotator drum | |
Accessory for Digital Cel-Gro Tissue Culture Rotator, 120V (88882015) and Digital Cel-Gro Tissue Culture Rotator 230V (88882016) |