missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Drums for Digital Cel-Gro Tissue Culture Rotators

Accessory for use with Digital Cel-Gro Tissue Culture Rotator, 120V and Digital Cel-Gro Tissue Culture Rotator, 230V

Brand:  Thermo Scientific™ 88882155

Additional Details : Weight : 1.95000kg

 View more versions of this product

Product Code. 17338282

  • £291.71 / Each
Estimated Shipment: 30-10-2024
to see stock.
Add to basket

Description

Description

Accessory for use with Digital Cel-Gro Tissue Culture Rotator, 120V and Digital Cel-Gro Tissue Culture Rotator, 230V

Specifications

Specifications

Rotating Drum
380 mm
diameter 17 mm tube
142
Stainless Steel
142 x diameter 17 mm tube rotator drum
Accessory for Digital Cel-Gro Tissue Culture Rotator, 120V (88882015) and Digital Cel-Gro Tissue Culture Rotator 230V (88882016)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers