Learn More
Thermo Scientific™ Dry-Bags™ Half Fraser Broth
Simplify media preparation in food laboratories with this high quality dehydrated medium in a convenient bag format; just open-fill-dispense, saving precious time in the lab.
Brand: Thermo Scientific™ DB0895L
2081.23 GBP valid until 2024-12-31
Use promo code "23263" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Save time in preparing bulk media and diluents for food microbiology testing using Thermo Scientific™ Dry-Bags™ Half Fraser Broth. The irradiated media is supplied in lightweight, transparent plastic bags — just add water and the medium is ready for use. Each bag makes 20L of medium, which can be dispensed through the outlet tube.
- Quick and easy to prepare — just add deionized water through filter sterilization to each Dry-Bag, and they are ready to use in minutes. Pre-sterilized water can also be added under aseptic conditions.
- Light to transport — there is no weighing of media or sterilization required, reducing the burden on media preparation departments and equipment.
- No handling of glassware or weighing of culture media required, no need to add supplementary reagents — empty bags can be disposed of with normal laboratory waste.
- Quality certificates are available for every batch to help reduce the burden of QC testing.
- Dry-Bags are easily adaptable to laboratory workflows and equipment such as automated diluters.
- Dry-Bags are easy to fill and dispense with minimum training.
- Conveniently packed in flat boxes of five or ten, Dry-Bags take up relatively little space compared to prepared media and can be made up as required.
Specifications
Clear, yellow | |
Dry-Bags Half Fraser Broth | |
Dehydrated Culture Media | |
20L |
10°C to 30°C | |
Dry-Bags™, with filter | |
5/Pk. |