missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Grant Instruments™ Dry Block Heating System, BTD Series
Click to view available options
Temperature Range (Metric):
Ambient +5° to 100°C
Description
The Grant BTD holds a combination of four microtube sizes simultaneously, and rapid heat-up enables swift change of application for maximum flexibility. The aluminum block is housed in a compact, stylish plastic moulding, which is durable, sturdy and easy to clean.
- Digital temperature control for optimum precision
- Heating range ambient + 5°C to 100°C
- Powerful heater for rapid heat-up times - 25°C to 100°C in just 15 minutes, 25°C to 37°C in just 4 minutes
- Stability ± 0.1°C, uniformity ± 0.2°C
- Heating block holds combinations of four microtube sizes simultaneously, up to a total of 49 tubes: 24 x 1.5/2.0mL, 15 x 0.5mL, 10 x 0.2mL
- Flexible tube sizes and rapid heat-up time enables swift change of application
- Convenient integral timer for time-sensitive incubations
- Sturdy, durable, easy-to-clean plastic construction; compact design with small footprint
- Two-line display for simple and precise setting of temperature/time and for monitoring current status during operation

Specifications
Specifications
Temperature Range (Metric) | Ambient +5° to 100°C |
Type | Dry Bath Block |
For Use With (Application) | Life-science/cancer research - DNA extraction incubations, DNA denaturation, PCR, ELISA and Western blotting, molecular biology |
Temperature Stability | 0.1°C |
Temperature Uniformity | ±0.1°C |
Timer | 1 minute to 96 hours (increments of 1 minute) |
Dimensions (L x W x H) | 230 x 210 x 110 mm |
Length (Metric) | 230 mm |
Height (Metric) | 115 mm |
Weight (Metric) | 2.8 kg |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Grant Instruments™ Dry Block Heating System, BTD Series >
Spot an opportunity for improvement?Share a Content Correction