missing translation for 'onlineSavingsMsg'
Learn More

Gardner Denver Welch™ Dry Ice/Isopropanol Trap (-79°C)

Effective foreline cold trap for vacuum manifolds or Schlenk Lines.

Brand:  Gardner Denver Welch™ U1420H-14

Product Code. 15321461

  • £1450.00 / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Recommended for vacuum manifold (Schlenk Line) use
  • Uses dry ice or liquid nitrogen as refrigerant
  • Removable center make trapping surface easy to clean
  • Has large 3 quart (2.8L) center well for the dry ice/isopropyl alcohol slurry or liquid nitrogen
  • Transparent ring allows visual monitoring of the trapping surface without breaking the vacuum while the system is operating
  • With dry ice cold temperature may be maintained for up to 12 hours depending upon the vapor load
  • With liquid nitrogen cold temperature can be maintained for up to 2 hours depending upon the vapor load
Specifications

Specifications

Vacuum manifold (Schlenk Line)
Dry Ice/Isopropanol Trap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.