missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Dry-Keeper Small, Stacking Gas-Purge Desiccator Cabinet

Space saving dry storage unit can be stacked 5 units high for maximum use of bench space.

Brand:  Bel-Art™ SP Scienceware™ H42053-0002

Product Code. 11342104

  • £524.00 / Each
Estimated Shipment: 14-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Pull down door slides out of way into compartment beneath cabinet
  • Rear mounted gas ports are tapered to accept tubing with0.25 to 0.375 in. I.D.
  • Interior volume: 0.1 cu. ft.
  • Gas ports can be closed to use desiccator with standard desiccant cartridges, 10555044, 11317194, 11867913
Specifications

Specifications

Not Autoclavable
Vertical
Pull-down
8.25 in.
10.62 in.
6.75 in.
Clear
Polystyrene
0.1 cu.ft.
21 cm
27 cm
17 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ SP Scienceware™ Dry-Keeper Small, Stacking Gas-Purge Desiccator Cabinet > Dimensions (L x W x H): 21 x 27 x 17cm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.