missing translation for 'onlineSavingsMsg'
Learn More

Welch Ilmvac™ Diaphragm Vacuum Pump Service and Maintenance Kit

Product Code. p-8012710 Shop All Welch Ilmvac Products
Click to view available options
For Use With:
MP/MPC 101 Z, 201 E
MP/MPC 201 T, MP 101V
MP/MPC 301 Z, 601 E, 301 Z
MP/MPC 601 T, 901 Z, 1201 E, MP 301V
MP/MPR 060 E, 030 Z, Biovac 060, MP 101V
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 13191984

Brand: Welch Ilmvac™ 402008

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Service and Maintenance Kits comprised of replacement seals and wearable parts for various models of diaphragm pumps.

  • Available for a range of diaphragm vacuum pump models
  • Provided individually
TRUSTED_SUSTAINABILITY

Specifications

For Use With MP/MPC 101 Z, 201 E
Includes Diaphragm, Valves, and O-rings
Quantity 1 Ea.
Type Service Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.