missing translation for 'onlineSavingsMsg'
Learn More

Welch™ WOB-L Piston Dry Vacuum Pump 2511

Perfect for many common laboratory vacuum applications. Welch™ WOB-L Piston Dry Vacuum Pump 2511 is reliable and economical.

Brand:  Welch™ U2511C-02

703.91 GBP valid until 2024-12-15
Use promo code "21658" to get your promotional price.


Product Code. 13492059

  • £1102.40 / Each
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Perfect for many common laboratory vacuum applications. Welch™ WOB-L Piston Dry Vacuum Pump 2511 is reliable and economical.

  • Economical
  • Oil free operation for reduced maintenance
Specifications

Specifications

21.6 in.Hg
194 x 114 x 191 mm
Dry Vacuum Pump
Common Laboratory Vacuum
230 V
230 V 50 Hz 1 Ph2.
European (Schuko)
11 L/min. at 60Hz
33 psig
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers