Learn More
BINDER™ Series ED Classic.Line - Drying and Heating Chambers, with Natural Convection
GREENER_CHOICE
The strengths of a BINDER ED series drying chamber include routine drying and sterilization tasks up to +300 °C. Thanks to the natural convection, all thermal processes which use this drying chamber are highly efficient. The ED series ensures fast, uniform drying.
Brand:  BINDER™ 9010-0075
 View more versions of this product
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 15602086
  • £4915.00 / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Uniform drying conditions thanks to APT.line™ technology
  • Identical test conditions throughout the chamber interior independent of sample size and quantity
  • Outstanding thermal insulation saves operation costs
  • Temperature range: room temperature plus 5 to 300°C
  • APT.line™ preheating chamber technology
  • Natural convection
  • Adjustable exhaust air flap
  • Controller with timer functions
  • 2 chrome-plated racks
  • Class 2 independent adjustable temperature safety device (DIN 12880) with visual alarm
Specifications
Specifications
ASTM D5374, DIN 12880
400 L
Drying and Heating Chamber
ED400-400 V-I
10/30/50/100 mm
150°C
3°C
1290 x 860 x 1050 mm
1025 mm
400 V
16 A
ED400-400 V-I, with RS 422, Series ED Classic.Line - Drying and heating chambers with natural convection, 400 L, 400 V 3 ∼ 50/60 Hz
5°C to 300°C
LED
10 (Maximum), 2 (Standard)
User friendly and easy to operate all-digital PID controller technology
0 to 99 hr
1000 x 520 x 800 mm
1235 mm
125 kg
3.4 kW
50/60 Hz
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
BINDER™ Series ED Classic.Line - Drying and Heating Chambers, with Natural Convection > Model: ED400-400V-I; Capacity: 400 L; Voltage: 400 V; Frequency: 50/60 Hz;