missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente Drying Tubes

Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.

Brand:  Lenz Laborglasintrumente 05339018

 View more versions of this product

Product Code. 10111621

  • £31.30 / Each
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Drying tubes
  • U-shaped Drying tubes
  • U-shaped
  • Side tubes with olives
  • Outer diameter 6 mm
  • Made of DURAN™ tubing
Specifications

Specifications

Olives
180 mm
U-shaped
Borosilicate Glass (3.3)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers