missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente Drying Tubes, U-Shaped, with NS Stopcocks

Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.

£103.00 - £114.00

Specifications

Material Borosilicate Glass (3.3)
View More Specs

Products 2
Product Code Brand Length (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Price Quantity & Availability  
10325691
View Documents
Lenz Laborglasintrumente
05342015
150 mm
£114.00
Each
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
 
10662472
View Documents
Lenz Laborglasintrumente
05342013
125 mm
£103.00
Each
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
 
Description

Description

  • Drying tubes acc. to DIN 12616
  • With removable NS stopcocks
  • Side tubes plain (outer dia.: 6 mm)
  • Made of DURAN™ tubing
Specifications

Specifications

Borosilicate Glass (3.3)
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lenz Laborglasintrumente Drying Tubes, U-Shaped, with NS Stopcocks

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.