Learn More
Kimberly-Clark™ KIMTECH™ White WETTASK DS Wipers
Controls costs and improves safety by reducing spills in busy healthcare environments. Kimberly-Clark™ KIMTECH™ White WETTASK DS Wipers provide a repeat use, airtight system using pre-soaked cloths.
Brand: Kimberly-Clark™ 7767
Code : 02
Additional Details : Weight : 0.01000kg
Description
- Perfect for infection control in busy healthcare environments
- Resistant to mould and bacteria
- Designed for use with a refillable, airtight wiping system using pre-soaked cloths to help control the costs of solvents and disinfectants and improve safety by reducing spills
Specifications
DS Wiper Roll | |
30.5 cm | |
31.80 x 30.50 cm |
31.8 cm | |
Fiber | |
Cleaning and disinfectant |