missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ dsbG (Escherichia coli) Recombinant Protein
Used for SDS-PAGE
Brand: Abnova™ P4977.100ug
This item is not returnable.
View return policy
Description
Sequence: MEELPAPVKAIEKQGITIIKTFDAPGGMKGYLGKYQDMGVTIYLTPDGKHAISGYMYNEKGENLSNTLIEKEIYAPAGREMWQRMEQSHWLLDGKPVIVYVFADPFCPYCKQFWQQARPWVDSGKVQLRTLLVGVIKPESPATAAAILASKDPAKTWQQYEASGGKLKLNVPANVSTEQMKVLSDNEKLMDDLGANVTPAIYYMSKENTLQQAVGLPDQKTLNIIMGNKSpecifications
NP_415137 | |
SDS-PAGE | |
945224 | |
dsbG (Escherichia coli) Recombinant protein | |
Conventional Chromatography | |
100 μg | |
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ECK0598/JW0597/ybdP | |
dsbG | |
None | |
Liquid |
1 mg/mL | |
Liquid | |
55.7kDa | |
Escherichia coli expression system | |
15% SDS-PAGE Stained with Coomassie Blue | |
MEELPAPVKAIEKQGITIIKTFDAPGGMKGYLGKYQDMGVTIYLTPDGKHAISGYMYNEKGENLSNTLIEKEIYAPAGREMWQRMEQSHWLLDGKPVIVYVFADPFCPYCKQFWQQARPWVDSGKVQLRTLLVGVIKPESPATAAAILASKDPAKTWQQYEASGGKLKLNVPANVSTEQMKVLSDNEKLMDDLGANVTPAIYYMSKENTLQQAVGLPDQKTLNIIMGNK | |
RUO | |
dsbG | |
E. coli | |
Escherichia coli expression system | |
>95% by SDS-PAGE |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction