missing translation for 'onlineSavingsMsg'
Learn More

IKA™ DT-20 Tubes with Rotor Stator Element

Comprised of tube, dispersing device, and cover. IKA™ DT-20 Tubes with Rotor Stator Element are used for dispersion, homogenisation, and suspensions with capacities ranging from 5 to 15mL.

Brand:  IKA™ 0003703100

Product Code. 10763304

  • £210.00 / Pack of 25
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Components: Tube, dispersing device, cover
Volume: 5 to 15mL
Application: Dispersion, homogenisation, suspensions
Additional media quantities may be added during operation sampling during operation
Packing unit: 25 pieces
Specifications

Specifications

DT-20 Tube
Dispersion, homogenization, suspensions
25 Pk.
5 to 15 mL
DT-20
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.