Learn More
Fisherbrand™ Anodized Dual Aluminum Blocks
Useful for a variety of applications in molecular biology, histology, clinical, environmental and industrial. Fisherbrand™ Anodized Dual Aluminum Blocks are for use with Fisherbrand Digital Block Heaters.
Brand:  Fisherbrand™ D1296-PCR
 View more versions of this product
Product Code. 11411997
  • £239.00 / Each
Estimated Shipment: 27-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Excellent heat transfer
  • Anodized to resist thermal shock
  • For holding test tubes for digital heaters
  • Available to fit a range of tube counts and sizes
Specifications
Specifications
Micro plate or 4 Slides.
D1196, 96-Well
1 CT