Learn More
Faster™ Dual Exhaust Internal HEPA Filter for SF/E SF/C CT/E 212 Cabinets
Dual Exhaust H14 HEPA Filter, built into the cabinet.
Brand:  Faster™ FX0021241000
Additional Details : Weight : 225.00000kg
Product Code. 16298670
  • £1100.00 / Each
Estimated Shipment: 25-09-2024
to see stock.
Description
Description
This accessory creates twin exhaust HEPA filters. Both exhaust HEPA filters are housed inside the main motor plenum. This has three main advantages:
1. The filter is inside the motor plenum and not in an external box. If the filter fails, it remains in negative pressure and therefore there is no risk of contamination leaving the cabinet.
2. The filter can be changed from the front of the cabinet, so no access is required at the top of the cabinet, which may not be available due to ceiling height.
3. The filter is inside the motor plenum therefore not adding height to the cabinet. This allows up to an 880 mm work surface height, with an external (floor to ceiling) height of 2345 mm (we recommend 100 to 200 mm space above the cabinet to ensure the correct level of airflow), providing a more conformable seating position for taller users. This still allows space at the top of the unit to fix and remove a fumigation top box.
Specifications
Specifications
H14 Hepa