missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Dual-Purpose Pipets

Dual-purpose pipets

Brand:  Fisherbrand™ 1138233G

 View more versions of this product

Product Code. 16660093

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Description

Description

  • Borosilicate glass, permanent amber markings, Tuf-Temp™-treated to reduce chipping
  • Accuracy printed on each pipet
  • Capacity marked below mouthpiece

Order Info

Available as PersonaPet™ personalized pipets

Compliance

Calibrated To Deliver/Blow Out (upper graduation) and To Contain (lower graduation).

Specifications

Specifications

±0.006 mL
0.06 oz.
Dual Purpose Pipet
Borosilicate Glass
2 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Dual-Purpose Pipets > Capacity: 2mL; Accuracy: ±0.006mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.