missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Dulcitol, 97%

£17.99 - £205.00

Chemical Identifiers

CAS 608-66-2
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.172
MDL Number MFCD00064288
InChI Key FBPFZTCFMRRESA-GUCUJZIJSA-N
Synonym dulcitol, galactitol, dulcite, d-galactitol, dulcose, euonymit, melampyrin, melampyrit, d-dulcitol, 2r,3s,4r,5s-hexane-1,2,3,4,5,6-hexaol
PubChem CID 11850
ChEBI CHEBI:16813
IUPAC Name (2R,3S,4R,5S)-hexane-1,2,3,4,5,6-hexol
SMILES C(C(C(C(C(CO)O)O)O)O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11418337
View Documents Promotion Details
Thermo Scientific Chemicals
A18402.14
25 g
£17.99
25g
Please to purchase this item. Need a web account? Register with us today!
11428337
View Documents Promotion Details
Thermo Scientific Chemicals
A18402.22
100 g
£50.80
100g
Please to purchase this item. Need a web account? Register with us today!
11438337
View Documents Promotion Details
Thermo Scientific Chemicals
A18402.36
500 g
£205.00
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Dulcitol is important in studies related to cataract formation, hepatosplenμly and mental retardation. It is used as a substrate to identify, differentiate and characterize galactitol dehydrogenase(s). It is also used as a reference compound in analytical procedures developed to analyze sugar alcohols in plants and food.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dulcitol is important in studies related to cataract formation, hepatosplenµly and mental retardation. It is used as a substrate to identify, differentiate and characterize galactitol dehydrogenase(s). It is also used as a reference compound in analytical procedures developed to analyze sugar alcohols in plants and food.

Solubility
Soluble in water (30g/L at 14°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is hygroscopic in nature.
Specifications

Chemical Identifiers

608-66-2
182.172
FBPFZTCFMRRESA-GUCUJZIJSA-N
11850
(2R,3S,4R,5S)-hexane-1,2,3,4,5,6-hexol
C6H14O6
MFCD00064288
dulcitol, galactitol, dulcite, d-galactitol, dulcose, euonymit, melampyrin, melampyrit, d-dulcitol, 2r,3s,4r,5s-hexane-1,2,3,4,5,6-hexaol
CHEBI:16813
C(C(C(C(C(CO)O)O)O)O)O

Specifications

608-66-2
1.47
C6H14O6
1721903
14,4332
Soluble in water (30g/L at 14°C).
C(C(C(C(C(CO)O)O)O)O)O
182.172
CHEBI:16813
97%
187°C to 191°C
275°C to 280°C (1 mmHg)
MFCD00064288
Hygroscopic
dulcitol, galactitol, dulcite, d-galactitol, dulcose, euonymit, melampyrin, melampyrit, d-dulcitol, 2r,3s,4r,5s-hexane-1,2,3,4,5,6-hexaol
FBPFZTCFMRRESA-GUCUJZIJSA-N
(2R,3S,4R,5S)-hexane-1,2,3,4,5,6-hexol
11850
182.17
Dulcitol
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 210-165-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only