Learn More
Macherey-Nagel™ Duotest™ pH Paper Roll
Featuring two indicator zones for more accurate readings. Macherey Nagel™ Duotest™ pH Paper Rolls give the user the best possible accuracy in pH measurement.
Brand:  Macherey-Nagel™ 90306
 View more versions of this product
Product Code. 10094880
  • £13.85 / 5m
Explore more special offers
Description
Description
Featuring two indicator zones for more accurate readings. Macherey Nagel™ Duotest™ pH Paper Rolls give the user the best possible accuracy in pH measurement.
  • Provides rapid and reliable results
  • Available for a range of pH gradations
  • Shows two different colours for each pH-value at intervals of 0.3 to 1 pH unit
  • Allows more accurate reading and good estimation of intermediate values
Specifications
Specifications
9.5 to 14
9.5, 10.0, 10.5, 11.0, 11.5, 12.0, 12.5, 13.0, 13.5, 14.0
pH Paper
10 mm
Roll
1 Each
5 m