missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- 12.00 pH value
- Immune to micro-organisms
- Expiration date on the bottle
- First class certificate with traceability to international standards
Specifications
Specifications
pH | 12.00 |
Physical Form | Liquid |
Quantity | 500 mL |
Solution Type | Buffer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DuraCal™ 12.00 Value pH Buffer, Hamilton™ >
Spot an opportunity for improvement?Share a Content Correction