missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuraCal™ Buffer pH 4.01, Hamilton™
Click to view available options
Quantity:
250 mL
Specifica
Specifica
Color | Red |
pH | 4.01 |
Physical Form | Liquid |
Quantity | 250 mL |
Solution Type | Buffer |
Solubility Information | Water Soluble |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'