missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuraCal™ Buffer pH 7, Hamilton™
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
Specifications
Specifications
| Color | Green |
| pH | 7 |
| Physical Form | Liquid |
| Quantity | 250 mL |
| Solution Type | Buffer |
| Solubility Information | Water Soluble |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction