missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuraCal™ 8.00 Value pH Buffer, Hamilton™
Click to view available options
Quantity:
500 mL
Unit Size:
Each
Description
- 8.00 pH value
- Immune to micro-organisms
- Expiration date on the bottle
- First class certificate with traceability to international standards
Specifications
Specifications
| pH | 8.00 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Solution Type | Buffer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction