missing translation for 'onlineSavingsMsg'
Learn More
DuraCal pH buffer 9.21, Hamilton™ Product Code.: 11567714

DuraCal pH buffer 9.21, Hamilton™

Product Code. 11567714
3 x500 mL
Click to view available options
Quantity:
3 x500 mL
This item is not returnable. View return policy

Product Code. 11567714

Brand: Hamilton 238919

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Prevent the growth of fungus and micro-organisms with this pH buffer. Hamilton™ DuraCal buffer 9.21 offers high buffering capacity and rapid, stable calibration.

  • Convenient 500mL bottle with built-in calibration compartment
  • Remains stable even in air
  • Economical, only about 15mL of buffer is used per calibration
  • Certified pH value from a DAkkS laboratory accredited for pH measurement
  • First class certificate with traceability to international standards
  • Certificates available here
  • Expiration date on the bottle
  • Immune to microbial growth

Specifications

pH 9.21
Quantity 3 x500 mL
Solubility Information 60Months
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DuraCal pH buffer 9.21, Hamilton™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.