Learn More
Lenz Laborglasinstrumente™ Condensers Acc. to Allihn
Facilitates the condensing of vapors in the laboratory. Lenz Laborglasinstrumente™ Condensers Acc. to Allihn are manufactured from durable DURAN™ borosilicate glass.
Brand: Lenz Laborglasinstrumente™ 05223211
Code : A0
Additional Details : Weight : 0.25000kg
Description
- Acc. to DIN 12581
- With NS socket and cone
- Corrosion-free and resistant to thermal shock
- Available with either glass olive or GL plastic olives/threads
Specifications
14/23 | |
Allihn Condenser | |
1 Pack |
For Condensing Vapors | |
160 mm | |
Duran Glass |