Learn More
Lenz Laborglasinstrumente™ Condensers Acc. to Allihn
Facilitates the condensing of vapors in the laboratory. Lenz Laborglasinstrumente™ Condensers Acc. to Allihn are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 05223214
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10743605
  • £82.00 / Each
Estimated Shipment: 06-09-2024
to see stock.
Add to basket
Description
Description
  • Acc. to DIN 12581
  • With NS socket and cone
  • Corrosion-free and resistant to thermal shock
  • Available with either glass olive or GL plastic olives/threads
Specifications
Specifications
29/32
Allihn Condenser
1 Pack
For Condensing Vapors
160 mm
Duran Glass