Learn More
Lenz Laborglasinstrumente™ Duran™ Allihn Condensers with Cone
Product Code. 10090201
Click to view available options
Includes:
100 - 250 mL Extractor
70mL Extractor
DURAN™ Olive Tubing
Unit Size:
Each
Product Code. 10090201
Brand: Lenz Laborglasinstrumente™ 05226006
Only null left
Add to basket
Add to basket
Specifications
For Use With (Application) For Condensing Vapors
Includes DURAN™ Olive Tubing
Quantity 1 Pack
Standard Taper Joint No NS 45/40
Material Duran Glass
Type Allihn Condenser