missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ ANSI/SLAS Microplate Holder
A microplate sized holder that allow small DURAN™ GL 25 bottles to be used with liquid handling robots
Brand: DWK Life Sciences
Additional Details : Weight : 0.20500kg
Description
- Liquid handling robots and pipetting systems are commonly used in the automation of chemical or biochemical laboratories.
- The systems automate a diverse range of the workflows used in drug discovery, genomics, proteomics and cell biology research.
- The containers used in the systems include microplates, tube racks and reservoirs.
- Allow the small 10 or 25 mL GL 25 DURAN™ bottles to be used on the decks of many liquid handing robots.
- Used in stability testing of new drug substances as analogues for the larger DURAN™ bottles. The small screw topped DURAN™ bottles are a robust, thicker-walled alternative to vials made from glass tubing.
- Convenient – Microplate holders meet the Footprint Dimensions in the ANSI/SBS standards
- Precise fit - Custom made for six 10 or 25 mL clear or amber DURAN GL 25 bottles in a 2 x 3 arrangement
- High quality - made of aluminum, milled for accuracy ''Made in Germany''
- Flexible usage - Aluminum has high thermal conductivity, so can be used with microplate heating or cooling systems
- Cost effective - Robust and reusable construction for a long service life
- Full traceability - Engraved lot-specific code for certification
- Future proof - An open space under the bottles allows computer vision technology to be used to identify presence of the bottles in the holder
Specifications
Microplate Holder for GL 25 Bottles | |
25 mL GL 25 Laboratory Bottles, 2 x 3 10 mL GL 25 Laboratory Bottles | |
127.76 mm | |
85.48 mm |
Bioassay Development, Primary Packaging Stability Studies, Automated Sample Testing, On Deck Reagent Reservoirs, Chemical Screening, Liquid Handling Robots | |
23 mm | |
361 g |