missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ ANSI/SLAS Microplate Holder
GREENER_CHOICE

Product Code. 16671076 Shop All DWK Life Sciences Products
Click to view available options
Weight (Metric):
296 g
361 g
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16671076

Brand: DWK Life Sciences 292331404

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A microplate sized holder that allow small DURAN™ GL 25 bottles to be used with liquid handling robots

  • Liquid handling robots and pipetting systems are commonly used in the automation of chemical or biochemical laboratories.
  • The systems automate a diverse range of the workflows used in drug discovery, genomics, proteomics and cell biology research.
  • The containers used in the systems include microplates, tube racks and reservoirs.
  • Allow the small 10 or 25 mL GL 25 DURAN™ bottles to be used on the decks of many liquid handing robots.
  • Used in stability testing of new drug substances as analogues for the larger DURAN™ bottles. The small screw topped DURAN™ bottles are a robust, thicker-walled alternative to vials made from glass tubing.
  • Convenient – Microplate holders meet the Footprint Dimensions in the ANSI/SBS standards
  • Precise fit - Custom made for six 10 or 25 mL clear or amber DURAN GL 25 bottles in a 2 x 3 arrangement
  • High quality - made of aluminum, milled for accuracy ''Made in Germany''
  • Flexible usage - Aluminum has high thermal conductivity, so can be used with microplate heating or cooling systems
  • Cost effective - Robust and reusable construction for a long service life
  • Full traceability - Engraved lot-specific code for certification
  • Future proof - An open space under the bottles allows computer vision technology to be used to identify presence of the bottles in the holder
TRUSTED_SUSTAINABILITY

Specifications

Description Microplate Holder for GL 25 Bottles
For Use With (Application) Bioassay Development, Primary Packaging Stability Studies, Automated Sample Testing, On Deck Reagent Reservoirs, Chemical Screening
For Use With (Equipment) 25 mL GL 25 Laboratory Bottles, 2 x 3 10 mL GL 25 Laboratory Bottles
Height (Metric) 23 mm
Length (Metric) 127.76 mm
Weight (Metric) 296 g
Width (Metric) 85.48 mm
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt