missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Aspirator Laboratory Bottle, Standard Taper Socket Side Arm, complete with Glass Stopcock and Glass Stopper
Click to view available options
Capacity (Metric):
1000 mL
2000 mL
500 mL
Description
Aspirator bottles, complete with glass stopcock. Complying with ISO 4796, these aspirators have a ground joint neck and glass stopper
- Supplied ready to use
- DURAN™ glass stopcock
- DURAN™ glass stopper
- Large range of sizes available
Specifications
Specifications
Capacity (Metric) | 2000 mL |
For Use With (Application) | Storage and dispensing of liquids and reagents |
Material | Borosilicate Glass (3.3) |
No. per Pack | 1/Pk. |
Neck Finish | Standard Taper Joint Bottom 19/26 |
Autoclavable | Autoclavable |
Diameter (Metric) | 134 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction