missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Aspirator Laboratory Bottle, Standard Taper Socket Side Arm, complete with Glass Stopcock and Glass Stopper
Aspirator bottle, complete with glass stopcock and glass stopper.
£191.00 - £1280.00
Specifications
For Use With (Application) | Storage and dispensing of liquids and reagents |
---|---|
Material | Borosilicate Glass (3.3) |
Autoclavable | Autoclavable |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15261999
|
DWK Life Sciences
247025401 |
1000 mL | N/A | ||||||
10097550
|
DWK Life Sciences
247026303 |
2000 mL |
£191.00
Each |
Estimated Shipment: 20-12-2024
Log in to see stock. |
|||||
15251999
|
DWK Life Sciences
247024405 |
500 mL | N/A | ||||||
Description
Aspirator bottles, complete with glass stopcock. Complying with ISO 4796, these aspirators have a ground joint neck and glass stopper
- Supplied ready to use
- DURAN™ glass stopcock
- DURAN™ glass stopper
- Large range of sizes available
Specifications
Storage and dispensing of liquids and reagents | |
Borosilicate Glass (3.3) | |
Autoclavable |