missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Blanking Screw Cap, GL 14, Red PBT, with PTFE Cap Liner (GL 45 Connection Cap System Accessory)
4.96 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Use promo code "21743" to get your promotional price.
Click to view available options
Red
Description
Flexible modular GL 45 connection cap system that accepts four different rigid or semi-rigid tubing diameters (1.6 mm; 3.0 mm; 3.2 mm and 6.0 mm). The caps are available with 2 or 3 GL 14 threaded ports. The different sized inserts form a tight seal around the pass-through tubing. Sterile pressure equalisation is possible through use of 0.2 micron syringe filter. Unused ports can be sealed off with a red GL 14 screw cap.
- Rigid or semi-rigid tubing of different diameters can be accepted by using a blue GL 14 open topped cap with one of the inserts
- Available with 2, or 3 threaded outlets (ports)
- Five different tubing diameters (3.2 mm, 6.0 mm, 8.0 mm, 10.0 mm and 12.0 mm) can be used by changing the inserts
- By applying positive or negative pressure liquid can be pumped out of or into the bottle via the screw cap
- Fully compatible with all the GL 45 threads and therefore highly versatile
- Fully compatible with all the GLS 80 threads and therefore highly versatile
Suitable Accessories:
- 1129750
- 1129751
Specifications
Diameter (Metric) | 87 mm |
Description | Screw cap,PBT, with PTFE cap liner, red, GL 14 |
For Use With (Application) | Sterile fluid transfer in and out of lab bottles; Gas transfer in and out of lab bottles; Fluid transfer between lab bottles; Supply of reagents to instruments; Bioreactor supply |
Material | Polypropylene, PTFE |
Quantity | 2/Pk. |
Thread Size (Metric) | GL 14 |
Autoclavable | Autoclavable |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'