Learn More
Lenz Laborglasinstrumente™ DURAN™ Borosilicate Glass Round Bottom Flasks with Conical Ground Joint
Brand: Lenz Laborglasinstrumente™ 03002988
This item is not returnable.
View return policy
Specifications
According to DIN EN ISO 4797 and in addition to standard | |
Borosilicate Glass, Duran Glass | |
Round |
10 L | |
29/32 NS Socket | |
279 mm |
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ DURAN™ Borosilicate Glass Round Bottom Flasks with Conical Ground Joint > Capacity/Socket Joint: 10000mL-29/32 NS Socket
Spot an opportunity for improvement?Share a Content Correction